Learn More
Eisco™ Visual Scientifics Atwood Machine Module Set
SCI_ED
Catalog No. S07812 Shop All Eisco Products
Change view
Click to view available options
Atwood Machine module set
Catalog No.
S07812 Atwood Machine module set
1 options
Catalog No. S07812 Supplier Eisco™ Supplier No. PTAWM
Add to Cart
Add to Cart
Includes
Atwood machine, right angle clamp, 3.5cm long spring with a spring constant of 21N/m, Visual Scientifics post, one roll string
Requires
Spring scale 2.5N or force probe, hooked mass set, video recording device, Visual Scientifics base, Visual Scientifics back board, small pads
Includes activities to help students understand simple machines and the idea of tension in a string when constructing a free body diagram.
  • Students understand how exactly pulleys work and the effect of placing a spring between two pulleys
  • Includes 10+ page instruction booklet covering approximately two to three hours of class/lab material, 10+ Next Generation Science Standards Concepts and 10+ Common Core Standards
  • Every module includes comprehensive instructions to assist with lesson planning, student activities and experimental procedures
  • All labs and extensions feature recommended video analysis setups and guides
Specifications
Product Type Atwood Machine Module Set
For Use With (Application) Ideal of tension in a string when constructing a free body diagram
Includes Atwood machine, right angle clamp, 3.5cm l spring with a spring constant of 21n/m, visual scientifics post, 1 roll string, instructions, student activities, experimental procedures