Learn More
Description
Includes
Two banana plugs
Requires
Eisco Sensors Base Unit
- Range: -20 to 20V
- Resolution: 0.01V
- Max Sampling: 3,000 S/sec.
- Testing batteries and electronic circuits, measuring potential of chemical cells, studying electronic components
Specifications
Specifications
| Type | Sensor, Voltage |
| Includes | Two banana plugs |
| For Use With (Application) | Testing batteries and electronic circuits, measuring potential of chemical cells, studying electronic components |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
