Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Wheel and Axle Model
SCI_ED

Catalog No. S07827 Shop All Eisco Products
Click to view available options
:
Wheel and Axle Model

Includes

Activity guide

Learn how this simple machine reduces the force needed to move a load.

  • Observe how a crane works
  • Learn about multiple uses for a wheel and axle
  • Durable wooden construction is ideal for hands-on classroom use

Specifications

Product Type Model, Wheel and Axle
Material Wood
For Use With (Application) Observe how a crane works
Includes Activity guide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.