Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                ELA3A Antibody, Novus Biologicals™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | ELA3A | 
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 | 
| Applications | Immunohistochemistry (Paraffin) | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
Description
ELA3A Polyclonal specifically detects ELA3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ELA3A | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| chymotrypsin-like elastase family member 3A, chymotrypsin-like elastase family, member 3A, EC 3.4.21, EC 3.4.21.70, ELA3A, ELA3elastase 1, elastase 3A, pancreatic, elastase 3A, pancreatic (protease E), Elastase IIIA, elastase-3A, Protease E | |
| CELA3A | |
| IgG | |
| Affinity Purified | 
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 10136 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:LFVHPLWNRSCVACGNDIALIKLSRSAQLGDAVQLASL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            