Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Antibody (CELA3B/1218), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$246.61 - $542.38

Specifications

Antigen Elastase
Clone CELA3B/1218
Concentration 0.2 mg/mL
Dilution Western Blot, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Applications Western Blot, Flow Cytometry, SDS-Page, Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24481200
SDP
View Documents
Novus Biologicals
NBP2448120.02MG
0.02 mg
Each for $246.61
Only null left
Add to Cart
 
NBP24481201
SDP
View Documents
Novus Biologicals
NBP2448120.1MG
0.1 mg
Each for $542.38
Only null left
Add to Cart
 
NBP24481202
SDP
View Documents
Novus Biologicals
NBP2448120.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunofluorescence

Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Elastase
0.2 mg/mL
Western Blot, Flow Cytometry, SDS-Page, Immunofluorescence
Unconjugated
Mouse
Cancer, Neuroscience
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
CELA3B
IgG κ
Protein A or G purified
This MAb recognizes a protein of ∼17kDa, identified as CELA3B (Chymotrypsin like elastase family member 3B). Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3B has little elastolytic activity. Like most of the human elastases, elastase 3B is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein; it has a digestive function in the intestine. Elastase 3B preferentially cleaves proteins after alanine residues. Elastase 3B may also function in the intestinal transport and metabolism of cholesterol. Both elastase 3A and elastase 3B have been referred to as protease E and as elastase 1, and excretion of this protein in fecal material is frequently used as a measure of pancreatic function in clinical assays.
CELA3B/1218
Western Blot, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human
P08861
23436
Recombinant human CELA3B protein
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.