Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Antibody (CELA3B/2810R), FITC, Novus Biologicals™
SDP

Catalog No. NB387378 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387378 Supplier Novus Biologicals Supplier No. NBP308372F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Elastase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CELA3B/2810R
Conjugate FITC
Formulation PBS
Gene Alias CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Host Species Rabbit
Immunogen Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary) (Uniprot: P08861)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23436
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.