Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Antibody (ELN/2069), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB383811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383811 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383811 Supplier Novus Biologicals Supplier No. NBP308889IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Elastin Monoclonal antibody specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Elastin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ELN/2069
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
Host Species Mouse
Immunogen Recombinant full-length human Elastin protein (Uniprot: P15502)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 2006
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.