Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastin Mouse anti-Human, Clone: ELN/2069, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30714720UG
Description
Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Elastin | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
Recombinant full-length human Elastin protein | |
20 μg | |
Extracellular Matrix | |
2006 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
ELN/2069 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction