Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Mouse anti-Human, Clone: ELN/2069, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Elastin
Clone ELN/2069
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121768
View Documents
Novus Biologicals
NBP30714720UG
20 μg
Each for $138.37
Add to cart
 
NB121767
View Documents
Novus Biologicals
NBP30714710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

Elastin
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
Recombinant full-length human Elastin protein
Monoclonal
Protein A or G purified
ELN/2069
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Extracellular Matrix
10 mM PBS with 0.05% BSA
2006
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit