Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELAVL4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15740220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15740220 20 μL
NBP157402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15740220 Supplier Novus Biologicals Supplier No. NBP15740220UL

Rabbit Polyclonal Antibody

ELAVL4 Polyclonal specifically detects ELAVL4 in Human samples. It is validated for Western Blot.

Specifications

Antigen ELAVL4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. P26378
Gene Alias abnormal vision, Drosophila, homolog of, like-4, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D), HuD, Paraneoplastic encephalomyelitis antigen HuD
Gene Symbols ELAVL4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ELAVL4(ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D)) The peptide sequence was selected from the N terminal of ELAVL4. Peptide sequence MQTGATTDDSKTNLIVNYLPQNMTQEEFRSLFGSIGEI
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1996
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.