Learn More
ELAVL4 Mouse, anti-Human, Unconjugated, Clone: 1B1F5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | ELAVL4 |
|---|---|
| Clone | 1B1F5 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9882
![]()
|
Proteintech Group Inc
678351IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ELAVL4, also named as HuD, is a 380 amino acid protein. The full name is neuronal embryonic lethal altered visual system-like(ELAVL) RNA-binding protein. ELAVL4 is expressed in brain and localized in the nucleus. PC12 cells expressing T7±HuD were treated with thio-specific (BMH) or amine-specific (DSS) chemical crosslinking reagents, and the resultant crosslinked complexes in the cell extracts were analyzed by immunoblotting with anti-T7 antibody. In addition to the monomeric HuD of 41 kDa, three specific crosslinked complexes could be detected using either of the crosslinkers. Two of the complexes had molecular masses of ∼80 and 120 kDa, and were likely to be the dimer and trimer of HuD, respectively. (PMID: 12384599 )
Specifications
| ELAVL4 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Rat, Mouse, Rabbit, Pig | |
| Primary |
| 1B1F5 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| 1996 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.