Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Electricity and Magnetism Kit
SDPSCI_ED

Catalog No. s13065 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
Catalog No. Class Size
S13065 1 to 2 Lab Groups
1 options

Catalog No. S13065

Supplier: United Scientific™ EMKIT3

Requires

Three AA batteries

The Electricity and Magnetism Kit is a comprehensive 29-piece kit that introduces students to the basic principles of electricity and magnetism, including magnetic attraction and repulsion magnetic fields, and electromagnetism.

  • Force in a magnetic field and electric motor basics are included, and light emitting diodes and a relay are provided for further investigations.
  • Components are mounted on sturdy molded bases and connections are made with reliable screw terminals.
  • An activity guide details the experiments.
  • Requires three AA batteries (not included).

Topics covered include:

  • Electrical circuitry
  • Voltage
  • Current
  • Resistance
  • Series and parallel connection

Specifications

Grade 5+
Electrical Requirements AA Batteries
For Use With (Application) Understanding principles of electricity and magnetism,
Includes Activity Guide
Quantity 29 Pieces/Kit
Requires Three AA batteries
Class Size 1 to 2 Lab Groups
Product Type Electricity and Magnetism Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.