Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Supplies Electrostatics Lab

Shop All United Scientific Supplies, Inc Products


Click to view available options
Class Size:
1 Lab Group
Description
- Demonstrates the following Electrostatic Principles: Electrostatic Attraction, Repulsion and Induction, Faraday’s Ice Pail Experiment and Surface Charge Distribution
Specifications
Specifications
| Grade | 9 to 15 |
| Age | 14 to 20 |
| Includes | Electroscopes with Ball Terminals, Disk Terminals for Electroscopes, Faraday Cage with Cover/Insulated Base, Aluminum Can, Glass Beaker, Electrophorus Plates/Handle, Charge Transfer Rods, Friction Rods (Polyethylene, PVC, Acrylic, Glass, Nylon, Polystyrene), Charge Transfer Ball, Pith Balls, Acrylic Sheet, Neon Bulb, Insulated Point; Acetate Cloth, and Activity Guide |
| Class Size | 1 Lab Group |
| Product Type | Electrostatics Lab |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction