Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF3/ESE-1 Antibody (SR1813), Novus Biologicals™
SDP

Catalog No. NB258162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258162 50 μg
NB259966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258162 Supplier Novus Biologicals Supplier No. NBP32229250UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ELF3/ESE-1 Monoclonal antibody specifically detects ELF3/ESE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen ELF3/ESE-1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1813
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR-1, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box
Host Species Rabbit
Immunogen A synthesized peptide derived from human ELF3/ESE-1 (Uniprot #: P78545)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1999
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.