Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF3/ESE-1 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB056247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056247 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056247 Supplier Novus Biologicals Supplier No. NBP298882H

Rabbit Polyclonal Antibody

ELF3/ESE-1 Polyclonal specifically detects ELF3/ESE-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ELF3/ESE-1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR-1, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ELF3/ESE-1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1999
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.