Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELFN1 Antibody (965705), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB339925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339925 Supplier Novus Biologicals Supplier No. FAB10644MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELFN1 Monoclonal specifically detects ELFN1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ELFN1
Applications Flow Cytometry
Classification Monoclonal
Clone 965705
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias USP17L7
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human ELFN1 protein, Asp28-Tyr418, Accession # P0C7U0
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392617
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.