Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics MGB Alert™™ Evaluation Sets

Primers, probes, and control sets are available in 48-reaction quantities for evaluation purposes

Specifications

Content And Storage 300μL Chemically synthesized DNA oligonucleotide primers
For Use With (Application) DNA Sequencing
No. of Reactions 48
View More Specs

Description

Description

Specifications

Specifications

300μL Chemically synthesized DNA oligonucleotide primers
DNA Sequencing
48
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.