Learn More
ELiTech Molecular Diagnostics Real Time PCR Positive Control Sets
Description
Each Positive Control ASR consists of plasmid containing the target insert associated with the relevant ASR primers and ASR probes product.
- Monitor aspects of PCR testing, lysis, extraction and PCR amplification, and detection
- Estimates testing precision and detects errors in laboratory testing procedures
- Effectively measures assay and instrument integrity
Specifications
Specifications
| Content And Storage | One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes |
| Kit Type | CMV 3 Positive Control Set |
| No. of Tests | 24 |
| For Use With (Application) | Real Time PCR (qPCR) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.