Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics Real Time PCR Positive Control Sets

Catalog No. 22800232
Change view
Click to view available options
BK Virus
C. pneumonia
CMV 3.0
EBV
Enterovirus
HHV-6
HSV-1
HSV-2
M. pneumonia
VZV
hMPV
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-800-232 EBV
22-800-231 CMV 3.0
22-800-230 BK Virus
22-800-238 VZV
22-800-239 HSV-1
22-800-233 HHV-6
22-800-237 M. pneumonia
22-800-236 C. pneumonia
22-800-234 Enterovirus
22-800-235 hMPV
22-800-240 HSV-2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-800-232 Supplier Nanogen, Inc Supplier No. M400393
This item is not returnable. View return policy

Verify proper performance of an assay and detect problems during the DNA or RNA extraction and amplification process associated with real-time PCR assay

Each Positive Control ASR consists of plasmid containing the target insert associated with the relevant ASR primers and ASR probes product.

  • Monitor aspects of PCR testing, lysis, extraction and PCR amplification, and detection
  • Estimates testing precision and detects errors in laboratory testing procedures
  • Effectively measures assay and instrument integrity

Specifications

Content And Storage One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes
Kit Type EBV Positive Control Set
No. of Tests 24
For Use With (Application) Real Time PCR (qPCR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.