Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1 Antibody (7E10D5), Novus Biologicals™
SDP

Catalog No. NB11060513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060513 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060513 Supplier Novus Biologicals Supplier No. NB11060513
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Elk-1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 7E10D5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Gene Symbols ELK1
Host Species Mouse
Immunogen Ni-NTA purified truncated recombinant ELK1 expressed in E. Coli strain BL21 (DE3)
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, MAP Kinase Signaling
Primary or Secondary Primary
Gene ID (Entrez) 2002
Test Specificity ELK1 (7E10D5)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.