Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1 Antibody (PCRP-ELK1-1D9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB389728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389728 Supplier Novus Biologicals Supplier No. NBP308888JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Elk-1 Monoclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen Elk-1
Applications Western Blot, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-ELK1-1D9
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Host Species Mouse
Immunogen Recombinant full-length human Elk-1 protein (Uniprot: P19419)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, MAP Kinase Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2002
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.