Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1, p Ser389 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343162 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343162 Supplier Novus Biologicals Supplier No. NBP254759MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Elk-1 Polyclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Elk-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Host Species Rabbit
Immunogen Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1.
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2002
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.