Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1, p Ser389 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254759 0.1 mg
NBP25475905 0.05 mg
NB413818 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP254759 Supplier Novus Biologicals Supplier No. NBP254759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Elk-1 Polyclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Elk1 (p Ser389)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Symbols ELK1
Host Species Rabbit
Immunogen Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1.
Molecular Weight of Antigen 62 kDa
Purification Method Epitope affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody detects endogenous level of Elk-1 only when phosphorylated at serine 389.
Target Species Human
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.