Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elk-1, p Ser389 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $682.00
Specifications
Concentration | 1 mg/mL |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Target Species | Human |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP254759
![]() |
Novus Biologicals
NBP254759 |
0.1 mg |
Each for $682.00
|
|
|||||
NBP25475905
![]() |
Novus Biologicals
NBP2547590.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB413818
![]() |
Novus Biologicals
NBP2547590.025MG |
0.025 mg |
Each for $196.10
|
|
|||||
Description
Elk-1 Polyclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
1 mg/mL | |
Unconjugated | |
Human | |
Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1. | |
Primary |
Polyclonal | |
Rabbit | |
ELK1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title