Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMO2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216318 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216318 Supplier Novus Biologicals Supplier No. NBP216318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELMO2 Polyclonal specifically detects ELMO2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ELMO2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96JJ3
Gene Alias ced-12 homolog 2, CED12A, ELMO-2, engulfment and cell motility 2, engulfment and cell motility 2 (ced-12 homolog, C. elegans), engulfment and cell motility protein 2, FLJ11656, hCed-12A, KIAA1834CED12CED-12, PH domain protein CED12A, Protein ced-12 homolog A
Gene Symbols ELMO2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ELMO2. The exact sequence is proprietary.
Molecular Weight of Antigen 83 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 63916
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.