Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMO2 Antibody, Novus Biologicals™
SDP

Catalog No. NB100879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100879 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100879 Supplier Novus Biologicals Supplier No. NB100879
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 5 publications

ELMO2 Polyclonal specifically detects ELMO2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen ELMO2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 3 - 6 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_877496.1
Gene Alias ced-12 homolog 2, CED12A, ELMO-2, engulfment and cell motility 2, engulfment and cell motility 2 (ced-12 homolog, C. elegans), engulfment and cell motility protein 2, FLJ11656, hCed-12A, KIAA1834CED12CED-12, PH domain protein CED12A, Protein ced-12 homolog A
Gene Symbols ELMO2
Host Species Goat
Immunogen Peptide with sequence C-PKEPSSYDFVYHYG corresponding to C-Terminus according to NP_877496.1, NP_573403.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 63916
Test Specificity NP_877496.1 and NP_573403.1 are varients that encode the same protein.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.