Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMSAN1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291003 Supplier Novus Biologicals Supplier No. NBP171912PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELMSAN1 Polyclonal antibody specifically detects ELMSAN1 in Human samples. It is validated for Western Blot, Immunoprecipitation, KnockDown

Specifications

Antigen ELMSAN1
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation, Knockdown Validated
Formulation PBS
Gene Alias c14_5541, C14orf117, C14orf43, chromosome 14 open reading frame 117, chromosome 14 open reading frame 43, ELM2 and Myb/SANT-like domain containing 1, ELMSAN1, FLJ00335, hypothetical protein LOC91748, LSR68
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 550 and 600 of human ELMSAN1 Open Reading Frame 43 using the numbering given in entry NP_001036783.1 (GeneID 91748).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91748
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.