Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL3 Antibody (V61 P2B3*D10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP2038A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2038A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2038A94 Supplier Novus Biologicals Supplier No. NBP250385AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELOVL3 Monoclonal specifically detects ELOVL3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen ELOVL3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V61 P2B3*D10
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3
Gene Symbols ELOVL3
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - QPYNFELSK
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 83401
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.