Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL5 Antibody (Z88), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP24809A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24809A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24809A30 Supplier Novus Biologicals Supplier No. NBP254809AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELOVL5 Monoclonal specifically detects ELOVL5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen ELOVL5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z88
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast)
Gene Symbols ELOVL5
Host Species Mouse
Immunogen Peptide Sequence NNVKPRKLR
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 60481
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.