Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL6 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342690 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342690 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342690 Supplier Novus Biologicals Supplier No. NBP281714MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELOVL6 Polyclonal antibody specifically detects ELOVL6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ELOVL6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast), ELOVL fatty acid elongase 6,3-keto acyl-CoA synthase ELOVL6, FAE, fatty acid elongase 2, long-chain fatty-acyl elongase
Host Species Rabbit
Immunogen ELOVL6 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of the human ELOVL6. The immunogen is located within the first 50 amino acids of ELOVL6. Amino Acid Squence: ENEAIQWMQENWKKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79071
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.