Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL7 Antibody (Z26), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309498 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309498 Supplier Novus Biologicals Supplier No. NBP250384MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELOVL7 Monoclonal antibody specifically detects ELOVL7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen ELOVL7
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z26
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias ELOVL family member 7, elongation of long chain fatty acids, ELOVL fatty acid elongase 7
Host Species Mouse
Immunogen Peptide Sequence - RAYTKGQRLP (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), bovine (Bos taurus), canine (Canis familiaris), equine (Equus caballus)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79993
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.