Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELYS Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB600238AFY Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600238AFY 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600238AFY Supplier Novus Biologicals Supplier No. NB600238AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELYS Polyclonal specifically detects ELYS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ELYS
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AT hook containing transcription factor 1, ELYS, ELYS transcription factor like protein TMBS62, ELYS transcription factor-like protein TMBS62, MST108, MSTP108, protein ELYS, putative AT-hook-containing transcription factor 1, TMBS62
Gene Symbols AHCTF1
Host Species Rabbit
Immunogen A syntetic peptide
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 25909
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.