Learn More
Description
Specifications
Specifications
| Antigen | ELYS |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AT hook containing transcription factor 1, ELYS, ELYS transcription factor like protein TMBS62, ELYS transcription factor-like protein TMBS62, MST108, MSTP108, protein ELYS, putative AT-hook-containing transcription factor 1, TMBS62 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ELYS fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
