Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMAP-II/AIMP1 Antibody (959507), Alexa Fluor 594, Novus Biologicals™
SDP

Catalog No. MB19101A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB19101A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB19101A594 Supplier Novus Biologicals Supplier No. MAB19101AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMAP-II/AIMP1 Monoclonal specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen EMAP-II/AIMP1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 959507
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Gene Symbols AIMP1
Host Species Mouse
Immunogen E.coli-derived recombinant human EMAP-II, Ser147-Lys312, Accession No. Q12904
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.