Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMAP-II/AIMP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227206 0.1 mg
NBP227206SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227206 Supplier Novus Biologicals Supplier No. NBP227206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

EMAP-II/AIMP1 Polyclonal specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EMAP-II/AIMP1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Gene Symbols AIMP1
Host Species Rabbit
Immunogen A portion of amino acids 720-800 from human AIMP1 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9255
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, orangutan, panda, cow, guinea pig, new world monkey, dog, Chinese hamster, horse, naked mole rat, rhesus monkey, opossum, rabbit, pig, cat, and white-cheeked gibbon, 94% in chimpanzee, frog, rat, platypus, wild turkey, elephant, chicken, and Carolina anole, and 83% homologous in zebrafish, cichlid fish, and catfish.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Other, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.