Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Embigin/EMB Antibody, Novus Biologicals™
SDP

Catalog No. NBP224689SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224689SS 0.025 mg
NBP224689 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224689SS Supplier Novus Biologicals Supplier No. NBP224689SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Embigin/EMB Polyclonal antibody specifically detects Embigin/EMB in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Embigin/EMB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25-2 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q6PCB8
Gene Alias emb, embigin, embigin homolog, embigin homolog (mouse), F9, GP70, H, MGC71745
Gene Symbols EMB
Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human Embigin was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133418
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.