Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Embigin/EMB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$160.07 - $406.62
Specifications
| Antigen | Embigin/EMB |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.25-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224689SS
![]() |
Novus Biologicals
NBP224689SS |
0.025 mg |
Each for $160.07
|
|
|||||
NBP224689
![]() |
Novus Biologicals
NBP224689 |
0.1 mg |
Each for $406.62
|
|
|||||
Description
Embigin/EMB Polyclonal antibody specifically detects Embigin/EMB in Human, Mouse samples. It is validated for Western Blot.Specifications
| Embigin/EMB | |
| Western Blot 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| emb, embigin, embigin homolog, embigin homolog (mouse), F9, GP70, H, MGC71745 | |
| EMB | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q6PCB8 | |
| 133418 | |
| A portion of amino acids 130-180 of human Embigin was used as the immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title