Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Protein A, Alkaline Phosphatase Conjugate
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
Description
- Validated for use in Direct Immunoassay
- Form: Liquid
- Formulation: In 50mM Tris, 1mM MgCl2, 1% BSA, pH 8.0
- Molar Mass: 50000
- Purity: Single band by SDS-PAGE
- Preservative: ≤0.1% sodium azide
- Applications: ELISA, Immunoblotting (Western Blotting)
Specifications
Specifications
| Product Type | Protein A, Alkaline Phosphatase Conjugate |
| Quantity | 1 mg |
| Content And Storage | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction