Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LiChrospher™ RP-18 ADS HPLC Cartridge

Catalog No. M1503850001 Shop All MilliporeSigma Products
Change view
Click to view available options
Length (Metric):
25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
M1503850001 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1503850001 Supplier MilliporeSigma™ Supplier No. 1.50385.0001

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates

  • The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography
  • Particles are 25μm diameter, spherical

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Length (Metric) 25 mm
Quantity 1/Pk.
Product Type LiChroCART Column/LiChrospher RP-18 ADS Sorbent HPLC Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.