Learn More
MilliporeSigma™ manu-CART™ Coupling Kits for LiChroCART™ Stainless-Steel Cartridges
Catalog No. p-4913508
Click to view available options
For Use With:
Coupling of 2 LiChroCART cartridges w/ same I.D.
Coupling of 25-4 or 25-2 cartridges
for capillary connections
TRUSTED_SUSTAINABILITY
Specifications
Type Coupling Kit
Packaging Quantity 1/Pk.
For Use With Coupling of 25-4 or 25-2 cartridges
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.