Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MultiScreen™ Sealing Tape
Shop All MilliporeSigma Products

Click to view available options
Color:
Clear
Description
- Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
- Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
- Precut to size
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Color | Clear |
Sterility | Non-sterile |
No. per Pack | 100 |
For Use With (Application) | Used to seal all manufacturers 300μL plate tops and plate bottoms for MicroBeta™ readers |
Product Type | Sealing Tape |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction