EMD Millipore Novagen™ BL21 Competent Cells Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 694494

 View more versions of this product

Catalog No. 69-449-4

  Check Availability
Add to Cart

    Specifications

    Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
    Contains F' Episome No
    Includes Blue/White Screening No
    Quantity 1mL
    Storage Requirements Store at ≤-70deg.C

    BL21 is the most widely used host background and has the advantage of being deficient in both lon (1) and ompT proteases.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit