Learn More
MilliporeSigma™ Novagen™ NovaBlue Singles™ Competent Cells
GSA_VA
Catalog No. 701813 Shop All MilliporeSigma Products
Click to view available options
Quantity:
11 Reactions
22 Reactions
Catalog No. 70-181-3
Supplier: MilliporeSigma™ 701813
Only null left
Add to Cart
Add to Cart
NovaBlue Singles Competent Cells are designed for ultimate convenience and reliability in plasmid transformation.
NovaBlue Singles Competent Cells, like all Novagen Singles Competent Cells, are designed for ultimate convenience and reliability in plasmid transformation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: >1.5 x 108 cfu/μg
Content And Storage ≤-70°C
Quantity 11 Reactions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.