Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Origami™ 2 (DE3) Competent Cells

Catalog No. p-7020096 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.4 mL
1 mL

Origami 2 strains have mutations in glutathione reducatase (gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation.

Origami™ 2 host strains are K-12 derivatives that have mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes, which greatly enhance disulfide bond formation in the E. coli cytoplasm. The Origami 2 strains are kanamycin sensitive; like the original Origami strains, the gor mutation is still selected for by tetracycline. To reduce the possibility of disulfide bond formation between molecules, strains containing mutations in trxB and gor are recommended only for the expression of proteins that require disulfide bond formation for proper folding. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.