Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Rosetta-gami™ 2 (DE3) pLacI Competent Cells

Catalog No. p-7020101 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.4 mL
1 mL

Rosetta-Gami 2 strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins. Contains the pLacI plasmid producing extra Lac repressor.

Expression host strains for pETBlue™ and pTriEx™ vectors are lDE3 lysogens that also carry the compatible pLacI plasmid, encoding the lac repressor. The pLacI plasmid confers chloramphenicol resistance and provides enough lac repressor to inhibit transcription from the T7lac promoter in the absence of inducer. The NovaBlue host strain is recommended for initial cloning with pETBlue and pTriEx vectors due to its superior transformation efficiency and high yields of high-quality plasmid DNA. Note that these strains are not recommended for use with pET, pACYCDuet™, pCDF, pRSF, pCOLADuet™ or pETcoco™ vectors. Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs. In Rosetta-gami 2(DE3)pLacI, the rare tRNA genes are present on the same plasmid that carries the lac repressor gene. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.