Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ RosettaBlue™ (DE3)pLysS Competent Cells

Catalog No. p-7020109 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.4 mL
1 mL

RosettaBlue host strains combine high transformation efficiency with enhanced expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.

RosettaBlue™ host strains are NovaBlue derivatives that combine high transformation efficiency and recA endA lacI&superscript_q; mutations with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, and GGA on a chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters. In RosettaBlue(DE3)pLysS, the rare tRNA genes are present on the same plasmid that carries the T7 lysozyme gene. Blue/white screening is not possible with RosettaBlue(DE3) strains due to the presence of the lacZ α-peptide coding sequence in the λDE3 prophage. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >1 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.