Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SeQuant™ ZIC™-cHILIC Metal-free Guard Column with One Coupler

Catalog No. M1506640001 Shop All MilliporeSigma Products
Change view
Click to view available options
Length (Metric):
20 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
M1506640001 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1506640001 Supplier MilliporeSigma™ Supplier No. 1.50664.0001

PEEK coated guard column 3 pc with coupler 1 pc

SeQuant™ ZIC™-cHILIC is designed for complementary selectivity and improved separation of polar hydrophilic compounds. This guard column protects your analytical separation column from contamination.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 20 mm
Quantity 1/Pk.
Product Type Guard Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.