Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Emerge™ Safety Eyewear
Suitable for both working environments and sports activities
$184.50 - $331.64
Specifications
Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
---|---|
Description | Will accommodate an Rx lens to the maximum of +3 to -3 diopters |
Magnification Power | None |
Weight (Metric) | 30 g |
Lens Material | Polycarbonate |
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
19-155-824
|
Pyramex™
SB7910DRX |
Black | Clear |
Case of 12 for $198.02
|
|
||||
19-155-825
|
Pyramex™
SB7950SF |
Black | IR/PC 5.0 |
Case of 12 for $331.64
|
|
||||
19-155-826
|
Pyramex™
SB7960SF |
Black | IR/PC 3.0 |
Case of 12 for $331.64
|
|
||||
19-155-938
|
Pyramex™
SG7910DRX |
Translucent Gray | Clear |
Case of 12 for $184.50
|
|
||||
Description
*NOTE: Emerge frame must be presented to your optometrist in order to be fitted with prescription lenses
Specifications
ANSI Z87.1,CSA Z94.3-07 | |
None | |
Polycarbonate |
Will accommodate an Rx lens to the maximum of +3 to -3 diopters | |
30 g | |
G300 Nylon |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title