Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMG1 Antibody (OTI1B8), PerCP, Novus Biologicals™
SDP

Catalog No. NB271601PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271601PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271601PCP Supplier Novus Biologicals Supplier No. NBP271601PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMG1 Monoclonal specifically detects EMG1 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen EMG1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1B8
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1, BWCNS, C2F18S rRNA Psi1248 methyltransferase, EMG1 nucleolar protein homolog (S. cerevisiae), essential for mitotic growth 1, FLJ60792, Grcc2f, NEP1EC 2.1.1.-, Nucleolar protein EMG1 homolog, probable ribosome biogenesis protein NEP1, Protein C2f, Ribosome biogenesis protein NEP1
Gene Symbols EMG1
Host Species Mouse
Immunogen Full length recombinant protein of human EMG1 (NP_006322) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10436.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.