Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMI1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177748 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177748 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177748 Supplier Novus Biologicals Supplier No. NBP177748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMI1 Polyclonal specifically detects EMI1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EMI1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.3 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:5000-1:20000, Immunohistochemistry 1:500-1:3000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9UK58
Gene Alias EMI1Early mitotic inhibitor 1, F-box only protein 5, F-box protein 5, FBX5F-box protein Fbx5, Fbxo31
Gene Symbols FBXO5
Host Species Rabbit
Immunogen This affinity purified EMI1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human EMI1 protein. (Uniprot: Q9UKT4)
Molecular Weight of Antigen 50.15 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26271
Test Specificity This antibody is directed against human Fbp5A protein. A BLAST analysis was used to suggest cross reactivity with Fbp5A protein.
Target Species Human, Primate
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.